siRNA Details
Gene Symbol: SOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003578.3 | NP_003569.1 | 2, 3 | 234 | ||
XM_011538849.2 | XP_011537151.1 | 2, 3 | 1287 | |||
XM_011538850.2 | XP_011537152.1 | 2, 3 | 1287 | |||
XM_011538851.2 | XP_011537153.1 | 2, 3 | 1287 | |||
GenBank mRNA | AF059203.1 | - | 2, 3 | 197 | ||
AF099031.2 | - | 2, 3 | 146 | |||
BC096090.1 | - | 2, 3 | 194 | 40000865 | ||
BC096091.3 | - | 2, 3 | 194 | 40000866 | ||
BC096092.3 | - | 2, 3 | 194 | 40000868 | ||
BC099626.3 | - | 2, 3 | 194 | 40000867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
