siRNA Details
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012576.2 | NP_036708.2 | 1 | 144 | ||
XM_008772065.1 | XP_008770287.1 | 1 | 145 | |||
XM_008772066.1 | XP_008770288.1 | 2 | 195 | |||
GenBank mRNA | AB115420.1 | - | 1 | 131 | ||
AF455050.1 | - | 1 | 138 | |||
AY066016.2 | - | 1 | 139 | |||
AY293740.1 | - | 1 | 92 | |||
M14053.1 | - | 1 | 199 | |||
Y00489.1 | - | 1 | 131 | |||
Y12264.1 | - | 1 | 153 |
