siRNA Details
Gene Symbol: Cacna1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012517.2 | NP_036649.2 | 15 | 2459 | ||
XM_006237159.2 | XP_006237221.1 | 16 | 2358 | |||
XM_006237170.2 | XP_006237232.1 | 15 | 2304 | |||
XM_006237171.2 | XP_006237233.1 | 15 | 2304 | |||
XM_006237172.2 | XP_006237234.1 | 15 | 2304 | |||
XM_006237175.2 | XP_006237237.1 | 16 | 2812 | |||
XM_008763216.1 | XP_008761438.1 | 16 | 4321 | |||
GenBank mRNA | AY974797.2 | - | 17 | 2576 | ||
DQ538522.1 | - | 17 | 2278 | |||
M59786.1 | - | 15 | 2462 | |||
M67515.1 | - | 15 | 2444 | |||
M67516.1 | - | 15 | 2938 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001159533, NM_001256001, NM_009781, NM_001159534, NM_001255997, and others.
Gene Ontology Categories:
