siRNA Details
Gene Symbol: CLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162407.1 | NP_001155879.1 | 4 | 885 | ||
NM_004071.3 | NP_004062.2 | 4 | 605 | |||
NR_027856.1 | 3 | 1627 | ||||
GenBank mRNA | AK025306.1 | - | 1 | 802 | ||
AK294295.1 | - | 4 | 885 | |||
AK307301.1 | - | 4 | 735 | |||
BC031549.1 | - | 4 | 501 | 4387232 | ||
L29219.1 | - | 4 | 579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
