siRNA Details
Gene Symbol: Hsd3b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012584.1 | NP_036716.1 | 1133 | |||
GenBank mRNA | BC092571.1 | - | 1150 | 7369915 | ||
M67465.1 | - | 1133 |
Gene Symbol: LOC100911116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749360.3 | XP_003749408.1 | 4 | 1195 |

More Information
Other Species Targeted

Gene Ontology Categories:
