siRNA Details
Gene Symbol: Tac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124770.1 | NP_001118242.1 | 4, 5 | 360 | ||
NM_012666.2 | NP_036798.1 | 4, 5 | 360 | |||
XM_008762692.1 | XP_008760914.1 | 4, 5 | 501 | |||
XM_008762694.1 | XP_008760916.1 | 4, 5 | 504 | |||
GenBank mRNA | M15191.1 | - | 4, 5 | 362 | ||
M34184.1 | - | 4, 5 | 362 |
