siRNA Details
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012680.3 | NP_036812.2 | 2 | 165 | ||
XM_006245909.2 | XP_006245971.1 | 2 | 346 | |||
XM_006245910.2 | XP_006245972.1 | 2 | 346 | |||
XM_006245911.2 | XP_006245973.1 | 2 | 346 | |||
XM_006245912.2 | XP_006245974.1 | 2 | 346 | |||
XM_006245913.2 | XP_006245975.1 | 2 | 346 | |||
XM_006245914.2 | XP_006245976.1 | 2 | 346 | |||
XM_006245915.2 | XP_006245977.1 | 2 | 346 | |||
XM_006245916.2 | XP_006245978.1 | 2 | 346 | |||
XM_006245917.2 | XP_006245979.1 | 2 | 346 | |||
XM_008767543.1 | XP_008765765.1 | 2 | 346 | |||
GenBank mRNA | D50413.1 | - | 1 | 117 | ||
U24150.1 | - | 2 | 165 |
