siRNA Details
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012687.1 | NP_036819.1 | 2 | 256 | ||
XM_008762759.1 | XP_008760981.1 | 2 | 317 | |||
XM_008762761.1 | XP_008760983.1 | 2 | 307 | |||
GenBank mRNA | D28773.1 | - | 2 | 256 | ||
D31798.1 | - | 2 | 178 |
