siRNA Details
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012696.2 | NP_036828.2 | 643 | |||
GenBank mRNA | BC087028.1 | - | 643 | 7130388 | ||
FQ211035.1 | - | 49 | ||||
FQ211199.1 | - | 49 | ||||
FQ219322.1 | - | 524 | ||||
FQ219531.1 | - | 49 | ||||
FQ219669.1 | - | 49 | ||||
M11883.1 | - | 413 | ||||
X02299.1 | - | 378 |
Gene Symbol: Kng1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009628.1 | NP_001009628.1 | 3, 4 | 487 | ||
GenBank mRNA | BC088161.1 | - | 3, 4 | 487 | 7369906 | |
FQ211311.1 | - | 1, 2 | 49 | |||
FQ219207.1 | - | 1, 2 | 49 | |||
FQ219641.1 | - | 1, 2 | 49 | |||
M11661.1 | - | 3, 4 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
