siRNA Details
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012596.1 | NP_036728.1 | 3 | 392 | ||
GenBank mRNA | AF287268.1 | - | 2 | 318 | ||
D84125.1 | - | 2 | 318 | |||
D84126.1 | - | 2 | 318 | |||
D84550.1 | - | 2 | 318 | |||
D84551.1 | - | 2 | 318 | |||
D85558.1 | - | 2 | 338 | |||
U52966.1 | - | 3 | 392 | |||
U53144.1 | - | 3 | 435 | |||
U60151.1 | - | 2 | 318 |
