siRNA Details
Gene Symbol: Myc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012603.2 | NP_036735.2 | 2 | 727 | ||
GenBank mRNA | AY679729.1 | - | 2 | 564 | ||
AY679730.1 | - | 2 | 727 | |||
BC091699.1 | - | 2 | 533 | 7303974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
