siRNA Details
Gene Symbol: Ucp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012682.2 | NP_036814.1 | 2 | 333 | ||
GenBank mRNA | BC088156.1 | - | 2 | 321 | 7380759 | |
M11814.1 | - | 2 | 331 | |||
X03894.1 | - | 2 | 243 |
