siRNA Details
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012690.2 | NP_036822.1 | 5 | 345 | ||
XM_006235999.2 | XP_006236061.1 | 3 | 405 | |||
XM_008762697.1 | XP_008760919.1 | 5 | 527 | |||
GenBank mRNA | L15079.1 | - | 5 | 347 |
