siRNA Details
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012696.2 | NP_036828.2 | 648 | |||
GenBank mRNA | BC087028.1 | - | 648 | 7130388 | ||
FQ211035.1 | - | 54 | ||||
FQ211199.1 | - | 54 | ||||
FQ219322.1 | - | 529 | ||||
FQ219531.1 | - | 54 | ||||
FQ219669.1 | - | 54 | ||||
M11883.1 | - | 418 | ||||
X02299.1 | - | 383 |
Gene Symbol: Kng1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009628.1 | NP_001009628.1 | 4 | 492 | ||
GenBank mRNA | BC088161.1 | - | 4 | 492 | 7369906 | |
FQ211311.1 | - | 2 | 54 | |||
FQ219207.1 | - | 2 | 54 | |||
FQ219641.1 | - | 2 | 54 | |||
M11661.1 | - | 4 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
