siRNA Details
Gene Symbol: Pfkfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271063.1 | NP_001257992.1 | 5, 6 | 516 | ||
NM_001271064.1 | NP_001257993.1 | 4, 5 | 390 | |||
NM_012621.4 | NP_036753.4 | 5, 6 | 639 | |||
XM_006256790.2 | XP_006256852.1 | 4, 5 | 2549 | |||
XM_006256791.2 | XP_006256853.1 | 5, 6 | 724 | |||
XM_008773123.1 | XP_008771345.1 | 7, 8 | 690 | |||
XM_008773124.1 | XP_008771346.1 | 7, 8 | 686 | |||
XM_008773125.1 | XP_008771347.1 | 6, 7 | 564 | |||
GenBank mRNA | AY707861.1 | - | 5, 6 | 467 | ||
GQ422136.1 | - | 4, 5 | 370 | |||
X15579.1 | - | 5, 6 | 496 | |||
X15580.1 | - | 4, 5 | 1494 | |||
Y00702.1 | - | 5, 6 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
