siRNA Details
Gene Symbol: RT1-EC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012645.1 | NP_036777.1 | 3 | 379 | ||
GenBank mRNA | AF400159.1 | - | 3 | 379 | ||
AJ004887.1 | - | 2 | 298 | |||
AJ004888.1 | - | 2 | 298 | |||
AJ005024.1 | - | 2 | 298 | |||
AJ005025.1 | - | 2 | 298 | |||
AJ249701.1 | - | 3 | 298 | |||
AJ249703.1 | - | 2 | 304 | |||
BC061817.1 | - | 4 | 380 | 5598840 | ||
L26224.1 | - | 3 | 379 | |||
L40363.1 | - | 4 | 358 | |||
M24023.1 | - | 2 | 148 | |||
M31038.1 | - | 3 | 379 | |||
X90371.1 | - | 2 | 298 | |||
X99767.1 | - | 2 | 298 |
Gene Symbol: RT1-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U38971.1 | - | 4 | 361 |
Gene Symbol: RT1-CE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008845.1 | NP_001008845.1 | 3 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
