siRNA Details
Gene Symbol: Tat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012668.2 | NP_036800.1 | 4, 5 | 460 | ||
GenBank mRNA | BC089813.1 | - | 4, 5 | 460 | 7372826 | |
M18340.1 | - | 4, 5 | 513 | |||
X02741.1 | - | 4, 5 | 513 |
