siRNA Details
Gene Symbol: Tcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012670.1 | NP_036802.1 | 8 | 901 | ||
GenBank mRNA | D90345.1 | - | 8 | 901 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013686, AK150666, AK166828, AK152633, AK149611, and others.
Gene Ontology Categories:
