siRNA Details
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012690.2 | NP_036822.1 | 7 | 602 | ||
XM_006235999.2 | XP_006236061.1 | 5 | 662 | |||
XM_006236000.1 | XP_006236062.1 | 3 | 325 | |||
XM_008762697.1 | XP_008760919.1 | 7 | 784 | |||
GenBank mRNA | L15079.1 | - | 7 | 604 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008830, BC150687, J03398, AK050345.
Gene Ontology Categories:
