siRNA Details
Gene Symbol: Kng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102418.1 | NP_001095888.1 | 1, 2 | 319 | ||
NM_012741.1 | NP_036873.1 | 1, 2 | 220 | |||
GenBank mRNA | BC088155.1 | - | 1, 2 | 319 | 7369450 | |
L29428.1 | - | 1, 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
