siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012789.1 | NP_036921.1 | 7, 8 | 571 | ||
XM_008761879.1 | XP_008760101.1 | 8, 9 | 1080 | |||
GenBank mRNA | J02997.1 | - | 7, 8 | 511 | ||
J04591.1 | - | 7, 8 | 571 |
