siRNA Details
Gene Symbol: Prss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012729.1 | NP_036861.1 | 3 | 254 | ||
GenBank mRNA | V01274.1 | - | 3 | 231 |
Gene Symbol: LOC103690254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762925.1 | XP_008761147.1 | 3 | 254 |
Gene Symbol: LOC683849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749732.3 | XP_003749780.1 | 3 | 254 |
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004097.1 | NP_001004097.1 | 3 | 254 | ||
XM_008762901.1 | XP_008761123.1 | 1 | 74 |

More Information
Other Species Targeted

Gene Ontology Categories:
