siRNA Details
Gene Symbol: Fcer1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012724.4 | NP_036856.1 | 3 | 550 | ||
XM_008769780.1 | XP_008768002.1 | 2 | 1255 | |||
XM_008769781.1 | XP_008768003.1 | 2 | 1255 | |||
GenBank mRNA | J03606.1 | - | 3 | 550 | ||
M17153.1 | - | 3 | 554 | |||
M21622.1 | - | 3 | 613 |
