siRNA Details
Gene Symbol: Prss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012729.1 | NP_036861.1 | 5 | 728 | ||
GenBank mRNA | L00131.1 | - | 2 | 274 | ||
V01274.1 | - | 5 | 705 |
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004097.1 | NP_001004097.1 | 5 | 728 | ||
XM_008762901.1 | XP_008761123.1 | 3 | 548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
