siRNA Details
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012731.2 | NP_036863.1 | 18 | 3056 | ||
XM_008771425.1 | XP_008769647.1 | 19 | 3060 | |||
XM_008771426.1 | XP_008769648.1 | 19 | 2946 | |||
GenBank mRNA | M55291.1 | - | 18 | 3059 |
