siRNA Details
Gene Symbol: ZNF211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209847.1 | - | 1 | 38 |
Gene Symbol: ZNF134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003435.4 | NP_003426.3 | 3 | 686 | ||
GenBank mRNA | AK291763.1 | - | 3 | 655 | ||
BC011646.2 | - | 2 | 545 | |||
BC021280.2 | - | 2 | 530 | |||
BC112294.1 | - | 3 | 446 | 8327762 | ||
BC113410.1 | - | 3 | 443 | 8322462 | ||
U09412.1 | - | 3 | 628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
