siRNA Details
Gene Symbol: Plagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012760.3 | NP_036892.1 | 7 | 2052 | ||
GenBank mRNA | U72620.1 | - | 7 | 2052 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
