siRNA Details
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012820.1 | NP_036952.1 | 2, 3 | 327 | ||
XM_006253122.1 | XP_006253184.1 | 3, 4 | 987 | |||
XM_006253123.1 | XP_006253185.1 | 3, 4 | 528 | |||
XM_006253124.2 | XP_006253186.1 | 3, 4 | 447 | |||
XM_006253125.2 | XP_006253187.1 | 3, 4 | 607 | |||
XM_006253126.1 | XP_006253188.1 | 3, 4 | 453 | |||
XM_006253127.1 | XP_006253189.1 | 3, 4 | 418 | |||
XM_008771254.1 | XP_008769476.1 | 2, 3 | 353 | |||
GenBank mRNA | D90109.1 | - | 2, 3 | 327 | ||
FQ217275.1 | - | 3, 4 | 399 |
