siRNA Details
Gene Symbol: Adcy6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270785.1 | NP_001257714.1 | 2 | 258 | ||
GenBank mRNA | L01115.1 | - | 2 | 401 | ||
M96160.1 | - | 2 | 258 |

More Information
Other Species Targeted

Gene Ontology Categories:
