siRNA Details
Gene Symbol: Asic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034014.1 | NP_001029186.1 | 1 | 349 | ||
GenBank mRNA | U53211.1 | - | 1 | 349 |
Gene Symbol: LOC103694861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773803.1 | XP_008772025.1 | 1 | 793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
