siRNA Details
Gene Symbol: Alx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012921.1 | NP_037053.1 | 1, 2 | 684 | ||
XM_006241293.2 | XP_006241355.1 | 2, 3 | 496 | |||
GenBank mRNA | BC158591.1 | - | 1, 2 | 460 | 9023971 | |
L14018.1 | - | 1, 2 | 684 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172553, AK086731, BC052200, AK047846.
Gene Ontology Categories:
