siRNA Details
Gene Symbol: PTK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004103.4 | NP_004094.3 | 16 | 1575 | ||
NM_173174.2 | NP_775266.1 | 20 | 2130 | |||
NM_173175.2 | NP_775267.1 | 15 | 1477 | |||
NM_173176.2 | NP_775268.1 | 15 | 1562 | |||
XM_005273447.4 | XP_005273504.1 | 15 | 1490 | |||
XM_005273448.4 | XP_005273505.1 | 15 | 1539 | |||
XM_011544441.2 | XP_011542743.1 | 15 | 1500 | |||
XM_011544442.2 | XP_011542744.1 | 15 | 1500 | |||
XM_017013214.1 | XP_016868703.1 | 16 | 1565 | |||
XM_017013215.1 | XP_016868704.1 | 15 | 1461 | |||
XM_017013216.1 | XP_016868705.1 | 15 | 1500 | |||
GenBank mRNA | AB209085.1 | - | 15 | 1472 | ||
AK128371.1 | - | 7 | 908 | |||
BC036651.2 | - | 20 | 2130 | 4824966 | ||
BC042599.2 | - | 15 | 1507 | 4343040 | ||
L49207.1 | - | 15 | 1502 | |||
U33284.1 | - | 15 | 1429 | |||
U43522.1 | - | 16 | 1565 |

More Information
Other Species Targeted

Gene Ontology Categories:
