siRNA Details
Gene Symbol: Sult1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012883.1 | NP_037015.1 | 3 | 332 | ||
GenBank mRNA | M86758.1 | - | 3 | 332 | ||
S76489.1 | - | 3 | 231 | |||
U50204.1 | - | 2 | 201 |
Gene Symbol: ste2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007718.2 | NP_001007719.1 | 3 | 308 | ||
XM_006250793.2 | XP_006250855.1 | 4 | 599 | |||
XM_008770039.1 | XP_008768261.1 | 5 | 513 | |||
GenBank mRNA | BC088157.1 | - | 3 | 308 | 7323209 | |
S76490.1 | - | 3 | 231 | |||
U50205.1 | - | 2 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
