siRNA Details
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108802.1 | NP_001102272.1 | 8 | 2813 | ||
NM_012905.2 | NP_037037.1 | 3 | 301 | |||
XM_006245252.2 | XP_006245314.1 | 10 | 2775 | |||
XM_006245255.2 | XP_006245317.1 | 3 | 2571 | |||
XM_006245256.2 | XP_006245318.1 | 3 | 600 | |||
XM_008767267.1 | XP_008765489.1 | 8 | 2875 | |||
XM_008767268.1 | XP_008765490.1 | 8 | 2652 | |||
XM_008767269.1 | XP_008765491.1 | 9 | 3269 | |||
XM_008767270.1 | XP_008765492.1 | 8 | 2828 | |||
XM_008767271.1 | XP_008765493.1 | 8 | 2827 | |||
GenBank mRNA | EV763588.1 | - | 3 | 301 | ||
U57097.1 | - | 3 | 277 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
