siRNA Details
Gene Symbol: Sult1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012883.1 | NP_037015.1 | 3 | 352 | ||
GenBank mRNA | M86758.1 | - | 3 | 352 | ||
S76489.1 | - | 3 | 251 | |||
U50204.1 | - | 2 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
