siRNA Details
Gene Symbol: PTK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004103.4 | NP_004094.3 | 3 | 366 | ||
NM_173174.2 | NP_775266.1 | 7 | 921 | |||
NM_173175.2 | NP_775267.1 | 2 | 268 | |||
NM_173176.2 | NP_775268.1 | 2 | 353 | |||
XM_005273447.4 | XP_005273504.1 | 2 | 281 | |||
XM_005273448.4 | XP_005273505.1 | 2 | 330 | |||
XM_011544441.2 | XP_011542743.1 | 2 | 291 | |||
XM_011544442.2 | XP_011542744.1 | 2 | 330 | |||
XM_017013214.1 | XP_016868703.1 | 3 | 356 | |||
XM_017013215.1 | XP_016868704.1 | 2 | 291 | |||
XM_017013216.1 | XP_016868705.1 | 2 | 330 | |||
GenBank mRNA | AB209085.1 | - | 2 | 263 | ||
BC036651.2 | - | 7 | 921 | 4824966 | ||
BC042599.2 | - | 2 | 298 | 4343040 | ||
BF796371.1 | - | 2 | 309 | |||
D45853.1 | - | 3 | 356 | |||
DA773624.1 | - | 2 | 353 | |||
L49207.1 | - | 2 | 293 | |||
U33284.1 | - | 2 | 220 | |||
U43522.1 | - | 3 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
