siRNA Details
Gene Symbol: Asic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034014.1 | NP_001029186.1 | 1 | 581 | ||
GenBank mRNA | U53211.1 | - | 1 | 581 |
Gene Symbol: LOC103694861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773803.1 | XP_008772025.1 | 1 | 1025 |

More Information
Other Species Targeted

Gene Ontology Categories:
