siRNA Details
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108802.1 | NP_001102272.1 | 9 | 2995 | ||
NM_012905.2 | NP_037037.1 | 4 | 483 | |||
XM_006245252.2 | XP_006245314.1 | 11 | 2957 | |||
XM_006245255.2 | XP_006245317.1 | 4 | 2753 | |||
XM_006245256.2 | XP_006245318.1 | 4 | 782 | |||
XM_008767267.1 | XP_008765489.1 | 9 | 3057 | |||
XM_008767268.1 | XP_008765490.1 | 9 | 2834 | |||
XM_008767269.1 | XP_008765491.1 | 10 | 3451 | |||
XM_008767270.1 | XP_008765492.1 | 9 | 3010 | |||
XM_008767271.1 | XP_008765493.1 | 9 | 3009 | |||
GenBank mRNA | BC087690.1 | - | 1 | 121 | ||
EV763588.1 | - | 4 | 483 | |||
U57097.1 | - | 4 | 459 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
