siRNA Details
Gene Symbol: Hrh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012965.3 | NP_037097.3 | 1 | 536 | ||
XM_008771486.1 | XP_008769708.1 | 1 | 1843 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hrh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012965.3 | NP_037097.3 | 1 | 536 | ||
XM_008771486.1 | XP_008769708.1 | 1 | 1843 |
Set Membership: |
![]() |