siRNA Details
Gene Symbol: Il1rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167840.1 | NP_001161312.1 | 3 | 431 | ||
NM_012968.1 | NP_037100.1 | 3 | 350 | |||
XM_006248504.2 | XP_006248566.1 | 3 | 419 | |||
GenBank mRNA | BC097339.1 | - | 3 | 366 | 7442849 | |
GU123169.1 | - | 3 | 431 | |||
U48592.1 | - | 3 | 350 |
