siRNA Details
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 2 | 291 | ||
GenBank mRNA | AF065147.1 | - | 2 | 203 | ||
BC061531.1 | - | 2 | 295 | |||
M61874.1 | - | 2 | 291 | |||
M61875.1 | - | 2 | 291 | |||
U46957.1 | - | 2 | 203 | |||
U52179.1 | - | 2 | 202 | |||
U96138.1 | - | 2 | 291 |
