siRNA Details
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013030.1 | NP_037162.1 | 10, 11 | 1216 | ||
GenBank mRNA | AB013455.1 | - | 1, 2 | 737 | ||
BC078876.1 | - | 10, 11 | 1210 | 7109613 | ||
L13257.1 | - | 10, 11 | 1216 |
