siRNA Details
Gene Symbol: Fosl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013146.1 | NP_001013164.1 | 3 | 287 | ||
NM_012954.1 | NP_037086.1 | 4 | 542 | |||
GenBank mRNA | AY622611.1 | - | 4 | 539 | ||
U18913.1 | - | 4 | 542 | |||
U18982.1 | - | 3 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
