siRNA Details
Gene Symbol: Kcna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012970.3 | NP_037102.1 | 3 | 980 | ||
GenBank mRNA | J04731.1 | - | 3 | 980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
