siRNA Details
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012999.1 | NP_037131.1 | 7 | 951 | ||
GenBank mRNA | D31854.1 | - | 5 | 572 | ||
L31894.1 | - | 7 | 951 |

More Information
Other Species Targeted

Gene Ontology Categories:
