siRNA Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013133.1 | NP_037265.1 | 2 | 356 | ||
XM_006246378.2 | XP_006246440.1 | 2 | 508 | |||
GenBank mRNA | AJ310834.1 | - | 2 | 172 | ||
AJ310835.1 | - | 2 | 172 | |||
AY827463.1 | - | 2 | 172 | |||
D00833.1 | - | 2 | 356 | |||
X55246.1 | - | 2 | 260 | |||
Y00276.1 | - | 1 | 108 |
