siRNA Details
Gene Symbol: Tsc22d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109912.1 | NP_001103382.1 | 3 | 3297 | ||
NM_013043.2 | NP_037175.1 | 3 | 425 | |||
XM_006252339.1 | XP_006252401.1 | 4 | 3169 | |||
GenBank mRNA | BC059146.1 | - | 3 | 436 | ||
BC127547.1 | - | 3 | 417 | |||
L25785.1 | - | 3 | 418 |

More Information
Other Species Targeted

Gene Ontology Categories:
