siRNA Details
Gene Symbol: Otc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013078.1 | NP_037210.1 | 8 | 877 | ||
GenBank mRNA | BC088158.1 | - | 8 | 843 | 7325875 | |
FQ219657.1 | - | 8 | 840 | |||
K00001.1 | - | 8 | 863 | |||
K03040.1 | - | 8 | 835 | |||
M11266.1 | - | 8 | 877 | |||
X01976.1 | - | 8 | 863 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008769, M17030, AK050930, BC024893.
Gene Ontology Categories:
