siRNA Details
Gene Symbol: Rps26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013224.1 | NP_037356.1 | 2 | 127 | ||
GenBank mRNA | BC061561.1 | - | 2 | 131 | 6921494 | |
FQ210888.1 | - | 2 | 127 | |||
FQ211456.1 | - | 2 | 126 | |||
FQ216976.1 | - | 2 | 127 | |||
FQ217039.1 | - | 2 | 127 | |||
FQ217630.1 | - | 2 | 127 | |||
FQ220682.1 | - | 2 | 127 | |||
FQ220803.1 | - | 2 | 198 | |||
FQ221742.1 | - | 2 | 127 | |||
FQ221917.1 | - | 2 | 128 | |||
FQ222007.1 | - | 2 | 127 | |||
FQ222170.1 | - | 2 | 127 | |||
FQ222423.1 | - | 2 | 128 | |||
FQ222638.1 | - | 2 | 127 | |||
FQ223096.1 | - | 2 | 127 | |||
FQ223202.1 | - | 2 | 127 | |||
FQ223253.1 | - | 2 | 123 | |||
FQ223633.1 | - | 2 | 127 | |||
FQ224810.1 | - | 2 | 127 | |||
FQ229027.1 | - | 2 | 127 | |||
FQ233862.1 | - | 2 | 127 | |||
FQ233965.1 | - | 1 | 127 | |||
FQ234974.1 | - | 2 | 128 | |||
FQ235169.1 | - | 2 | 128 | |||
FQ235253.1 | - | 2 | 127 | |||
X02414.1 | - | 2 | 127 |
Gene Symbol: LOC100360508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728001.3 | XP_002728047.2 | 1 | 161 |
Gene Symbol: LOC100361756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730273.3 | XP_002730319.3 | 1 | 101 |
Gene Symbol: LOC100361854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730210.4 | XP_002730256.1 | 1 | 121 |
Gene Symbol: RGD1559808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236845.5 | XP_236845.1 | 1 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
