siRNA Details
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017008.4 | NP_058704.1 | 3 | 260 | ||
GenBank mRNA | AB017801.1 | - | 3 | 226 | ||
BC059110.1 | - | 3 | 261 | 6919611 | ||
BC087743.1 | - | 3 | 229 | 7321006 | ||
DQ403053.1 | - | 1 | 116 | |||
M17701.1 | - | 2 | 215 | |||
U75401.1 | - | 2 | 210 | |||
X02231.1 | - | 3 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
